Tisk…

Resazurin sodium salt, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin

M-Clarity™ quality level = MQ200

R7017-1G R7017-5G
SIALR7017-1GEA 1520 CZK
SIALR7017-1G SIALR7017-5G
Resazurin sodium salt, Sigma-Aldrich®
Resazurin sodium salt
Vzorec: C₁₂H₆NNaO₄
Storage Temperature: Ambient
MDL Number: MFCD00005036
CAS číslo: 62758-13-8
EINECS: 263-718-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR