Tisk…

Resazurin sodium salt, pure, high purity biological stain

Dodavatel: Thermo Fisher Scientific
Varování

189900010 189900050 189900250
ACRO189900010EA 1320 CZK
ACRO189900010 ACRO189900050 ACRO189900250
Resazurin sodium salt, pure, high purity biological stain
Resazurin sodium salt
Vzorec: C₁₂H₆NNaO₄
Storage Temperature: Ambient
MDL Number: MFCD00005036
CAS číslo: 62758-13-8
EINECS: 263-718-5

Objednat

Výsledky specifikačního testu

Appearance Dark green to black Powder
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥75 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR