Tisk…

Azomethine H sodium salt hydrate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Sodium 5-hydroxy-4-((2-hydroxybenzylidene)amino)-7-sulphonaphthalene-2-sulphonate xhydrate, Azomethine-H monosodium salt hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt hydrate, Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate hydrate, Azomethine-H sodium hydrate

11635-25G 11635-5G
SIAL11635-25GEA 5590 CZK
SIAL11635-25G SIAL11635-5G
Azomethine H sodium salt hydrate, Sigma-Aldrich®
Azomethine H sodium salt hydrate
Vzorec: C₁₇H₁₂NNaO₈S₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149588
CAS číslo: 206752-32-1
EINECS: 227-698-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR