Tisk…

Azomethine H sodium salt hydrate

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Sodium 5-hydroxy-4-((2-hydroxybenzylidene)amino)-7-sulphonaphthalene-2-sulphonate xhydrate, Azomethine-H monosodium salt hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt hydrate, Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate hydrate, Azomethine-H sodium hydrate

A10927.03 A10927.06 A10927.14
A10927.03EA 961 CZK
A10927.03 A10927.06 A10927.14
Azomethine H sodium salt hydrate
Azomethine H sodium salt hydrate

CAS No.: 206752-32-1 Molecular Formula: C17H12NNaO8S2.xH2O Formula Weight: 445.41(anhy) Melting Point: 300° Beilstein Reference: 4100175 MDL No.: MFCD00149588

Vzorec: C₁₇H₁₂NNaO₈S₂·xH₂O
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149588
CAS číslo: 206752-32-1
EINECS: 250-975-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR