Tisk…

Indigo carmine, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

M-Clarity™ quality level = MQ200

131164-100G 131164-25G
SIAL131164-100GEA 6730 CZK
SIAL131164-100G SIAL131164-25G
Indigo carmine, Sigma-Aldrich®
Indigo carmine
Vzorec: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36
Bod tání: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005723
CAS číslo: 860-22-0
EINECS: 212-728-8
Merck Index: 13,04969

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR