Indigo carmine, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue
57000-100G-F
57000-25G-F
57000-500G-F
SIAL57000-100G-FEA
5080
CZK
SIAL57000-100G-F
SIAL57000-25G-F
SIAL57000-500G-F
Indigo carmine, Sigma-Aldrich®
Indigo carmine
Vzorec:
C₁₆H₈N₂Na₂O₈S₂ MW: 466.36 Bod tání: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005723 CAS číslo: 860-22-0 EINECS: 212-728-8 Merck Index: 13,04969 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...