Tisk…

Docusate sodium ≥98%, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate

M-Clarity™ quality level = MQ200

323586-100G 323586-5G 323586-500G
SIAL323586-100GEA 2010 CZK
SIAL323586-100G SIAL323586-5G SIAL323586-500G
Docusate sodium ≥98%, Sigma-Aldrich®
Docusate sodium
Vzorec: C₂₀H₃₇NaO₇S
Bod varu: ∼180 °C (1013 hPa)
Bod tání: ∼155 °C
Hustota: ∼1,1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012455
CAS číslo: 577-11-7
EINECS: 209-406-4
Merck Index: 12,03460

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR