Tisk…

Bismarck brown Y

Dodavatel: Thermo Scientific

Synonyms: C.I. 21000, 1,3-Bis(2,3-diaminophenylazo)benzene dihydrochloride, Bismarck Brown Y (G), Basic Brown 1, Vesuvine, Phenylene Brown, Leather Brown, Basic Brown G, GX, GXP, Manchester Brown, 4,4'-[1,3-Phenylenebis(azo)]bisbenzene-1,3-diamine dihydrochloride

A16674.14 A16674.22
A16674.14EA 1220 CZK
A16674.14 A16674.22
Bismarck brown Y
Bismarck brown Y

CAS No.: 10114-58-6 Molecular Formula: C18H18N8.2HCl Formula Weight: 419.32 Odour: Odourless Solubility: Soluble in water Merck Reference: 14,1253 MDL No.: MFCD00012977


Plasma stain for mucins, cartilage and goblet cells

Vzorec: C₁₈H₁₈N8·2HCl
MW: 419,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012977
CAS číslo: 10114-58-6
EINECS: 233-314-3
Merck Index: 14,1253

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR