Bismarck brown R, black powder C.I. 21010
Dodavatel: MP Biomedicals
Synonyms:
C.I. 21010
0215485850
0215485880
ICNA0215485850EA
1310
CZK
ICNA0215485850
ICNA0215485880
Bismarck brown R, black powder C.I. 21010
Bismarck brown R
Bismarck Brown R is used in a modified Coomasie blue staining procedure.
Vzorec:
C₂₁H₂₄N₈·2HCl MW: 461,39 g/mol Bod tání: 222 °C (rozkl., lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00036386 CAS číslo: 5421-66-9 EINECS: 213-888-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...