Tisk…

Antimony(III) potassium oxitartrate hydrate ≥99.95% (trace metals basis), Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: (R,R)-(+)-Antimony potassium tartrate hydrate, (+)-Antimony potassium L-tartrate hydrate, L(+)-Tartaric acid antimony potassium salt hydrate, L(+)-Antimony potassium tartrate hydrate, Antimony potassium L(+)-tartrate hydrate, (R,R)-(+)-Tartaric acid antimony potassium salt hydrate, Antimony potassium tartrate hydrate, Potassium antimony(III) tartrate hydrate

M-Clarity™ quality level = MQ100

230057-100G 230057-10G
SIAL230057-100GEA 31910 CZK
SIAL230057-100G SIAL230057-10G
Antimony(III) potassium oxitartrate hydrate ≥99.95% (trace metals basis), Sigma-Aldrich®
Antimony(III) potassium oxitartrate hydrate
Vzorec: C₈H₄K₂O₁₂Sb₂·H₂O
MW: 631,84 g/mol (anhy)
MDL Number: MFCD00148863
CAS číslo: 331753-56-1
EINECS: 234-293-3
UN: 1551
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR