Tisk…

Antimony(III) potassium oxitartrate hydrate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (R,R)-(+)-Antimony potassium tartrate hydrate, (+)-Antimony potassium L-tartrate hydrate, L(+)-Tartaric acid antimony potassium salt hydrate, L(+)-Antimony potassium tartrate hydrate, Antimony potassium L(+)-tartrate hydrate, (R,R)-(+)-Tartaric acid antimony potassium salt hydrate, Antimony potassium tartrate hydrate, Potassium antimony(III) tartrate hydrate

A10889.22 A10889.0E A10889.36
A10889.22EA 1200 CZK
A10889.22 A10889.0E A10889.36
Antimony(III) potassium oxitartrate hydrate ≥98%
Antimony(III) potassium oxitartrate hydrate

Purity: 98% CAS No.: 331753-56-1 Molecular Formula: C8H4K2O12Sb2.xH2O Formula Weight: 613.82(anhy) Physical Form: Powder Density: 2.607 Appearance: White UN No.: 1551 MDL No.: MFCD00148863

Vzorec: C₈H₄K₂O₁₂Sb₂·H₂O
Hustota: 2,607
Storage Temperature: Ambient
MDL Number: MFCD00148863
CAS číslo: 331753-56-1
EINECS: 234-293-3
UN: 1551
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR