Tisk…

(-)-Quinine hydrochloride dihydrate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate

M-Clarity™ quality level = MQ200

8.22194.0025 8.22194.0100
8.22194.0025EA 1750 CZK
8.22194.0025 8.22194.0100
(-)-Quinine hydrochloride dihydrate, Sigma-Aldrich®
(-)-Quinine hydrochloride dihydrate
Vzorec: C₂₀H₂₅ClN₂O₂·2H₂O
MW: 396.91
Bod tání: 158…160 °C
Bod vzplanutí: 122
Storage Temperature: Ambient
MDL Number: MFCD00151248
CAS číslo: 6119-47-7
EINECS: 205-001-1

Objednat

Výsledky specifikačního testu

Solubility 62.5 g/l (20 °C)
Melting Point 115 °C (decomposition)
Molar Mass 396.91 g/mol
Bulk Density 310 kg/m3
Flash Point 122 °C
pH 6.0 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR