Quinine hemisulphate monohydrate ≥98.0%, BioReagent suitable for fluorescence, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Quinine sulphate, Quinine sulphate (2:1) (salt) dihydrate
M-Clarity™ quality level = MQ200
22640-25G
22640-5G
SIAL22640-25GEA
6240
CZK
SIAL22640-25G
SIAL22640-5G
Quinine hemisulphate monohydrate ≥98.0%, BioReagent suitable for fluorescence, Sigma-Aldrich®
Quinine hemisulphate monohydrate
Vzorec:
C₂₀H₂₄N₂O₂·0.5H₂SO₄·H₂O MW: 391.47 g/mol (373.46 anhy) Hustota: 1.36 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150792 CAS číslo: 6119-70-6 EINECS: 212-359-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...