(-)-Quinine, anhydrous 99% (total base) may cont. up to 5% dihydroquinine
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
A10459.09
A10459.18
A10459.30
A10459.09EA
1470
CZK
A10459.09
A10459.18
A10459.30
(-)-Quinine, anhydrous 99% (total base) may cont. up to 5% dihydroquinine
(-)-Quinine
Purity: 99%
CAS No.: 130-95-0
Molecular Formula: C20H24N2O2
Formula Weight: 324.42
Melting Point: ca 177° dec.
Beilstein Reference: 91867
Merck Reference: 14,8061
Optical Rotation: -166° (c=1 in ethanol)
MDL No.: MFCD00198096
Vzorec:
C₂₀H₂₄N₂O₂ MW: 324.42 Bod tání: ∼ 177 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00198096 CAS číslo: 130-95-0 EINECS: 205-003-2 Merck Index: 14,8061 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...