(-)-Cinchonidine 98.5-101% (dry basis)
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(8α,9R)-Cinchonan-9-ol
110330250
110331000
ACRO110330250EA
2550
CZK
ACRO110330250
ACRO110331000
(-)-Cinchonidine 98.5-101% (dry basis)
(-)-Cinchonidine
Vzorec:
C₁₉H₂₂N₂O MW: 294,4 g/mol Bod varu: 465 °C (1013 hPa) Bod tání: 200…203 °C Hustota: 1,22 g/cm³ (20 °C) |
MDL Number:
MFCD00006783 CAS číslo: 485-71-2 EINECS: 207-622-3 Merck Index: 13,02307 |
Výsledky specifikačního testu
Appearance | White to cream Crystalline powder |
Infrared spectrum | Conforms |
Melting point | ≥200 °C |
Loss on drying | ≤1 % (105 °C) |
Specific optical rotation | -105° to -115° (20 °C, 589 nm) (c=1, EtOH) (on dried substance) |
Titration with HClO4 | 98.5 to 101.0 % (On dried substance) |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...