Tisk…

(-)-Cinchonidine 99% (total base) may cont. up to 5% quinine

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (8α,9R)-Cinchonan-9-ol

A18796.14 A18796.22
A18796.14EA 2350 CZK
A18796.14 A18796.22
(-)-Cinchonidine 99% (total base) may cont. up to 5% quinine
(-)-Cinchonidine

Purity: 99% CAS No.: 485-71-2 Molecular Formula: C19H22N2O Formula Weight: 294.39 Melting Point: 201-206° Beilstein Reference: 89690 Merck Reference: 14,2286 Optical Rotation: -109° (c=0.5 in ethanol) MDL No.: MFCD00006783

Vzorec: C₁₉H₂₂N₂O
MW: 294,4 g/mol
Bod varu: 465 °C (1013 hPa)
Bod tání: 201…206 °C
Hustota: 1,22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006783
CAS číslo: 485-71-2
EINECS: 207-622-3
Merck Index: 14,2286

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR