D-(+)-Menthol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(1S)-(+)-Menthol, (1R,2S,4S)-(+)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1S,2R,5S)-(+)-Menthol, D(+)-Menthyl alcoholol
M-Clarity™ quality level = MQ100
224464-10G
224464-50G
SIAL224464-10GEA
2170
CZK
SIAL224464-10G
SIAL224464-50G
D-(+)-Menthol, Sigma-Aldrich®
D-(+)-Menthol
Vzorec:
C₁₀H₂₀O MW: 156,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00062983 CAS číslo: 15356-60-2 EINECS: 239-387-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...