(±)-Camphene (sum of camphene + fenchene), tech. (sum of camphene + fenchene)
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Camphene, (±)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane, (±)-3,3-Dimethyl-2-methylennorcamphane
A18040.30
A18040.18
A18040.0B
A18040.30EA
2260
CZK
A18040.30
A18040.18
A18040.0B
(±)-Camphene (sum of camphene + fenchene), tech. (sum of camphene + fenchene)
(±)-Camphene
CAS No.: 5794-04-7
Molecular Formula: C10H16
Formula Weight: 136.24
Flash Point: 34°(94°F)
Melting Point: ca 35°
Boiling Point: 159-160°
Merck Reference: 14,1730
UN No.: 1325
MDL No.: MFCD00066603
Vzorec:
C₁₀H₁₆ MW: 136,24 g/mol Bod varu: 159…160 °C Bod tání: 42…45 °C Hustota: 0,850 Bod vzplanutí: 34 °C (94 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00066603 CAS číslo: 79-92-5 EINECS: 201-234-8 UN: 1325 ADR: 4.1,III Merck Index: 14,1730 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...