Tisk…

L(-)-Menthol ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol

A10474.0B A10474.18 A10474.30
A10474.0BEA 9340 CZK
A10474.0B A10474.18 A10474.30
L(-)-Menthol ≥99%
L(-)-Menthol

Purity: 99% CAS No.: 2216-51-5 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 93°(199°F) Melting Point: 42-45° Boiling Point: 212-216° Beilstein Reference: 1902293 Merck Reference: 14,5837 Optical Rotation: -50° (c=10 in ethanol) MDL No.: MFCD00062979

Vzorec: C₁₀H₂₀O
MW: 156.27
Bod varu: 212…216 °C
Bod tání: 42…45 °C
Hustota: 0.89
Bod vzplanutí: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00062979
CAS číslo: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR