(±)-Camphene mixture of isomers for synthesis, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Camphene, (±)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane, (±)-3,3-Dimethyl-2-methylennorcamphane
M-Clarity™ quality level = MQ200
8.20254.0100
8.20254.1000
8.20254.0100EA
1010
CZK
8.20254.0100
8.20254.1000
(±)-Camphene mixture of isomers for synthesis, Sigma-Aldrich®
(±)-Camphene
Vzorec:
C₁₀H₁₆ MW: 136,24 g/mol Bod varu: 158…159 °C (1013 hPa) Bod tání: 48…52 °C Hustota: 0,85 g/cm³ (20 °C) Bod vzplanutí: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066603 CAS číslo: 79-92-5 EINECS: 201-234-8 UN: 1325 ADR: 4.1,II |
Výsledky specifikačního testu
Assay isomers (GC, area%) | ≥ 90.0 % (a/a) |
Assay (GC, Tricyclene) | ≤ 10.0 % (a/a) |
Identity (IR) | passes test |
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt. | 158 - 159 °C (1013 hPa) |
The product consists as a mixture of isomers >80,0 % camphene and >17,0 % fenchene. | 3.99 hPa (20 °C) |
Flash Point | 26 °C |
Density | 0.85 g/cm3 (20 °C) |
pH | 5.5 (H2O, 22 °C) (saturated aqueous solution) |
Ignition Point | 265 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...