Tisk…
Accessories for rotary evaporators
Dodavatel: Heidolph Instruments GmbH & Co.KG
569-00040-00 569-00030-00 569-00020-00
531-0844EA 7940 CZK
531-0844 531-0843 531-1068
Accessories for rotary evaporators
Odparky Odparky vakuové
Vapour temperature sensor: For advantage and precision models only - Vapour temperature display.
Vacuum sensor AUTOaccurate: For precision models with G3 and G6 glassware. The sensor allows automatic vacuum regulation for automatic solvent distillation and solvent mixing.
Vacuum accessories, vacuum valve: Required for all Hei-VAP models with valve regulated pumps.
- External construction designed for easy cleaning
- Can be combined with a Woulff bottle for one compact unit
Woulff bottle: Additional recovery of solvent in conjunction with condenser to protect vacuum valve and vacuum pump.
- Direct connection to the vacuum valve
- Features connection to vacuum controller
For Advantage and Precision models only.