Tisk…
Accessories and replacement parts
Dodavatel: VACUUBRAND
20696810 20696811 20696812 20696813 20696814 20696815 20696819 20696821 20696840 20696841 20699926 20637777
181-1671EA 1440 CZK
181-1671 181-1672 181-1673 181-1675 181-1676 181-1677 181-1674 181-1678 181-1621 181-1622 181-1653 VACU20637777
Accessories and replacement parts
Čerpadla Pumpy vakuové membránové
Versions available for pressure and vacuum fine tuning. Easy to assemble.
Ideal for solvent recovery. Emission condensers allow recovery rates up to almost 100% without costly refrigeration packages or cold traps. The AK + EK combination brings together the advantages of inlet separators, to prevent droplets and particles entering and damaging the pump, and promoting efficient solvent recovery.