Tisk…
Ultra-accurate digital thermometers, TFN 520 and 530
Dodavatel: EBRO
1340-5520 1340-5522 1340-5530 1340-5532
620-1460EA 6410 CZK
620-1460 620-1390 620-1391 620-1392
Ultra-accurate digital thermometers, TFN 520 and 530
Teploměry Teploměry digitální
These thermometers can be used for measuring temperatures in air conditioning and ventilation applications, air and gases, liquids and for measuring surface temperatures, temperatures of tools for injection moulding processes and plastic materials.
  • Maximum/minimum values displayed
  • SMP, Lemo or Ex proof connections
  • 'HOLD' function
  • Automatic switch-off
  • Response times (t99) of three seconds (620-1461)
External thermocouple types: K, J, T, E probes.
Standard version IP 52, SMP versions IP 40.
Weight: 90 g
Standard version IP 52, SMP versions IP 40.
Informace o dodávce: Supplied with three-point factory certificate of calibration, battery with charge indicator, without probe.