Tisk…
Vacuum Stirrer Shafts, Kombi-Vac
Dodavatel: GREINER GASSNER
6.0029.10 6.0034.10 6.0045.10 6.0029.16 6.0034.16 6.0045.16 6.0060.16 6.0074.16 6.1029.10 6.0034.00 6.1045.10 6.0060.00 6.0070.00 6.0000.10 6.0000.16 6.0001.10
441-9801EA 24240 CZK
441-9801 441-9802 441-9803 441-9804 441-9805 441-9806 441-9807 441-9808 441-9811 441-9812 441-9813 441-9814 441-9815 441-9821 441-9822 441-9831
Vacuum Stirrer Shafts, Kombi-Vac
Míchadla Hřídele míchací
Borosilicate glass with PTFE stirrer shaft insert and PTB screw cap ideal fit based on the screw fixing and insert position. The medium or gas to be stirred comes into contact with glass or PTFE. As vacuum-tight stirring cannot be achieved due to the cold flow of PTFE (Hostaflon), a sealing element made of Viton is located in the top part of the replaceable stirrer shaft. This sealing element creates a vacuum-tight connection between the stirrer shaft and agitator shafts. This design ensures excellent results from mixing trials in a vacuum, as well as for long-term trials at high speeds.
  • No power loss due to direct drive
  • Centred operation at high stirring speeds
  • All joint and flange sizes possible
  • Long service life; all stirrer shaft components can be replaced and reordered