Tisk…
Complete peristaltic pump sets
Dodavatel: Heidolph Instruments GmbH & Co.KG
523-51019-00 523-51019-02 523-51019-04
224-0033EA 114530 CZK
224-0033 224-1428 HEID523-51019-04
Complete peristaltic pump sets
Čerpadla Čerpadla peristaltická
Complete sets consisting of tube drive, pump head, and tubing. Ready-to-use.
  • Rated flow 0,3 to 80 ml/min
  • For tubing with 1,6 mm wall thickness
  • The tubing set includes 1 m Tygon tubing (internal diameter 0,8 mm) and 1 m silicone tubing (internal diameter 0,8 mm)
Peristaltic pump drive PD 5101 (for technical description see item 224-1352) with SP Quick 1,6 pump head and tubing set.