Tisk…
Drum pumps, ultra-pure
Dodavatel: BURKLE
5606-1000 5606-0600 5606-1001 5606-0601 5606-0400 5606-0401
223-0038EA 20310 CZK
223-0038 223-0094 223-0096 233-0121 223-0119 223-0120
Drum pumps, ultra-pure
Čerpadla Pumpy sudové
These pumps are ideal for decanting high purity liquids. Materials in contact with liquid are PTFE and FEP, these enable liquids, which attack conventional materials such as PP and PVC, to be pumped (except elemental fluorine). The stopcock, made of PTFE, prevents the escape of hazardous vapours and stops dripping.
  • Excellent chemical resistance
  • Threaded drum connections for commercially available containers are available
  • Infinitely variable adjustment of pump immersion depth guarantees good residual extraction
PP contact parts and flexible PVC tubing.