Tisk…
Watch glass dishes, DURAN®
Dodavatel: DWK Life Sciences
233214101 233216608 233212409 233213208 233213405 233213808 233214307 233214607 233215106 233215209 233215706 233216102
216-0277EA 917 CZK
216-0277 216-0283 SCOT233212409 SCOT233213208 SCOT233213405 216-0276 216-0278 SCOT233214607 216-0279 216-0280 216-0281 216-0282
Watch glass dishes, DURAN®
Skla hodinová
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fused edges.
Certifikace: DIN 12341