Tisk…
Watch glass dishes, DURAN®
Dodavatel: DWK Life Sciences
213213207 213213404 213214109 213214606 213215208 213215705 213216607 213212408 213216101
216-1112EA 552 CZK
216-1112 216-1114 216-1116 216-1118 216-1120 216-1122 216-1126 216-1110 216-1124
Watch glass dishes, DURAN®
Skla hodinová
DURAN®, borosilicate glass 3.3, clear, with fused edges.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
DIN 12341
Certifikace: DIN 12341