Tisk…
Certified screw top vials and closures
Dodavatel: Agilent
5182-0714 5182-0715 5182-0716
548-0762EA 945 CZK
548-0762 548-0763 548-0764
Certified screw top vials and closures
Vialky
These vials are certified for full warranted compatibility with Agilent autosamplers. The screw caps and precision fit septa give a secure seal with microvolume inserts.
  • Precision-formed neck for optimal robotic arm handling
  • 40% larger opening than standard narrow opening vials
  • Optional ceramic write-on spot with fill marks