Tisk…
Standard conical ground joints
Dodavatel: Lenz Laborglas GmbH & CO.KG
01100105 01100107 01100110 01100112 01100114 01100119 01100124 01100129 01100134 01100140 01100145 01100150 01100155 01100160 01100171 01100205 01100207 01100210 01100212 01100214 01100219 01100224 01100229 01100234 01100240 01100245 01100250 01100255 01100260 01100271 01100329 01100340 01100350 01100629 01100707 01100729 01101001 01101002 01100285 01100314 01100319 01100324 01100345 01101444 01100310 01100619 01100307 01100312 01100614 01100714 01101005 01101007 01101011 01101201 01101203 01100334 01100185 01100355 01100360 01100371 01100385 01100419 01100424 01100429 01100514 01100519 01100524 01100529 01100605 01100607 01100610 01100612 01100624 01100705 01100710 01100712 01100719 01100724 01100814 01100819 01100824 01100829 01101202 01101204 01101205 01101411 01101414 01101422
271-4101EA 472 CZK
271-4101 271-4102 201-4103 201-4104 201-4105 201-4106 201-4107 201-4108 271-4109 201-0934 201-4110 201-0935 201-0936 271-4111 201-0937 271-4131 271-4132 201-4133 201-4134 201-4135 201-4136 201-4137 201-4138 271-4139 201-0939 201-4140 201-0940 201-0941 271-4141 201-0942 271-4178 201-0945 201-0946 LENZ01100629 LENZ01100707 LENZ01100729 LENZ01101001 LENZ01101002 271-4113 271-4175 271-4176 271-4177 271-4160 LENZ01101444 201-0944 LENZ01100619 201-0943 271-4154 LENZ01100614 LENZ01100714 LENZ01101005 LENZ01101007 LENZ01101011 LENZ01101201 LENZ01101203 271-4159 201-0938 201-0947 271-4161 271-4162 201-0948 LENZ01100419 LENZ01100424 LENZ01100429 LENZ01100514 LENZ01100519 LENZ01100524 LENZ01100529 LENZ01100605 LENZ01100607 LENZ01100610 LENZ01100612 LENZ01100624 LENZ01100705 LENZ01100710 LENZ01100712 LENZ01100719 LENZ01100724 LENZ01100814 LENZ01100819 LENZ01100824 LENZ01100829 LENZ01101202 LENZ01101204 LENZ01101205 LENZ01101411 LENZ01101414 LENZ01101422
Standard conical ground joints
Adaptéry Adaptéry s kónickým zábrusů
DURAN®, borosilicate glass, clear.
  • Leakproof
  • Minimal requirement for joint lubrication
  • Joints cannot be misaligned
Socket and cone joints with fused edges. Excellent chemical resistance. High temperature resistance. Minimal thermal expansion giving high resistance to temperature changes.
DIN 12242 Form E (Socket joints)
DIN 12242 Form B (Cone joints)
ISO 383
Certifikace: DIN 12242 Form E (Socket joints), DIN 12242 Form B (Cone joints), ISO 383.