Tisk…
Tubing, Tygon® XL-60
Dodavatel: Saint Gobain Life Sciences
AN800003 AN800007 AN800012 AN800017 AN800022 AN800038 AN800046 AN800053
228-1056EA 8030 CZK
228-1056 228-1057 228-1058 228-1059 228-1060 228-1062 228-1063 228-1064
Tubing, Tygon® XL-60
Hadičky Přenosová hadička
Thermoplastic elastomer, translucent.
  • Excellent chemical resistance to many liquids, acids and bases
  • Resistant to ozone, sunlight and weathering
  • Extremely flexible and durable
Specially developed for use in peristaltic pumps, offering a service life of more than 500 hours when used in peristaltic pumps. The ideal replacement for PVC and silicone tubing when longer service life is required. Materials contain only small amounts of extractable substances, which reduces the risk of contamination during liquid transfer.
Temperature resistance: −40 to +120 °C
Hardness (Shore durometer A): 60
Density: 0,90
Certifikace: FDA 21 CFR 177.1210, NSF 51, EU Regulation 10/2011
Informace o dodávce: Shipped as rolls 15 m long in a cardboard box.