Tisk…
Screw caps for culture tubes, Corning®
Dodavatel: Corning
9999-132 9999-152 9999-24 9999-381M
212-0442EA 2770 CZK
212-0442 212-0443 212-0444 CORN9999-381M
Screw caps for culture tubes, Corning®
Uzávěry Víčka na zkumavky
These reusable Corning® caps are fabricated with specially formulated resin to resist the effects of temperature and steam in autoclaving.
For test, culture and centrifuge tubes with thread.