Tisk…
Čepicové proužky, plochý vršek
Dodavatel: Thermo Fisher Scientific
AB-0783 AB-0786 AB-0850 AB-0851 AB-0784
732-4880EA 996 CZK
732-4880 732-4882 732-4896 732-4897 732-4881
Čepicové proužky, plochý vršek
Těsnění Těsnění mikrodestičky
Made from polypropylene (PP).
  • Flat surface of each cap facilitates sample labelling
  • Large labelled end tabs for strip orientation and removal (A-H for 8 strip, 1 to 12 for 12 strip)
  • Certified free from DNase, RNase and Human DNA
Suitable for use with most of the 96-well PCR plates, as well as standard and low profile tube strips.
Colour: Natural