Tisk…
Centrifuge tubes, conical bottom
Dodavatel: Simport Scientific
T408-1 T408-2 T408
212-9607EA 6670 CZK
212-9607 212-9608 525-0387
Centrifuge tubes, conical bottom
Zkumavky Zkumavky centrifugační
PP or PS, conical bottom, graduated, disposable.
  • Suitable for general centrifugation, urinalysis procedures and serum separation
  • Chemically clean and metal free, ready to use and uniform in size and shape
  • Graduated
Max. RCF: 3000×g for PP tubes, 1200×g for PS tubes.