Tisk…
Centrifuge tubes, Nalgene®
Dodavatel: Thermo Fisher Scientific
3117-0120 3117-0150 3117-0160 3117-0380 3117-0500 3117-1000 3117-9500
525-2883EA 1280 CZK
525-2883 525-2884 525-2885 525-2887 525-2888 525-2890 525-2889
Centrifuge tubes, Nalgene®
Zkumavky Zkumavky centrifugační
PC, round bottom and rimless, autoclavable.
  • Transparent
  • Withstands forces up to 50000×g
  • Suitable for use in refrigerated or non refrigerated centrifuges
Informace pro objednávání: 525-2889 and 525-2890 are lipped.