Tisk…
Crucible tongs, stainless steel
Dodavatel: HAMMACHER KARL
HWN300-22 HWN301-25 HWN302-32
231-0274EA 2370 CZK
231-0274 231-0275 231-0276
Crucible tongs, stainless steel
Kleště
Stainless steel ¹⁸/₁₀, anti-magnetic. For easy, safe handling of crucibles made from a wide variety of materials.
  • Double bend
  • With grooved tips
  • Acid-resistant
  • Very good thermal resistance, autoclavable