Tisk…
Swabs with transport medium for viruses and Chlamydia
Dodavatel: Copan
144C 147C 148C 145C
710-0450EA 1780 CZK
710-0450 710-0453 710-0454 710-0451
Swabs with transport medium for viruses and Chlamydia
Tampóny odběrové Tampóny přepravní
Liquid transport medium is contained in a sponge carrier located in the bottom of the tube.
  • Special colours of caps to make differentiation easy
  • Simple insert swab into the tube after collecting sample
  • Swab tip placed in direct contact with the medium soaked sponge
Certifikace: ISO 13485. ISO 9001:2008.
Balení: Individually sealed in a peel-pouch. 25 swabs per box, 10x25 swabs per case.
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904