Tisk…
Držáky na kádinky a baňky, SP Bel-Art
Dodavatel: Bel-Art Products, a Part of SP
F183090100 F183090250
214-1935EA 7700 CZK
214-1935 214-1936
Držáky na kádinky a baňky, SP Bel-Art
Podstavce Podstavce pro baňky
Lead ring, with red vinyl coating and four stainless steel brackets. Holds beakers and flasks firmly in a fixed position. Ideal for use in water baths as it allows liquid to circulate beneath the vessel base.
  • Good chemical resistance
  • Good thermal resistance
  • Easy, simple handling