Tisk…
Stopcocks, three way
Dodavatel: Lenz Laborglas GmbH & CO.KG
02501401 02501402 02501802 02501804 02502404 02502906 02541401 02541402 02541802 02541804 02542404 02542906
227-3231EA 1190 CZK
227-3231 201-1035 205-2226 201-1036 205-2227 227-3235 201-1109 201-1192 201-3134 201-1037 201-3135 227-3215
Stopcocks, three way
Kohoutky
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys with T-bore
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
DIN 12554