Tisk…
Adhesive PCR film and foil seals
Dodavatel: Thermo Fisher Scientific
AB-0558 AB-0626
732-4819EA 4200 CZK
732-4819 732-4838
Adhesive PCR film and foil seals
Těsnění Těsnění mikrodestičky
Clear polyester film, aluminium foil. Recommended for use in PCR in conjunction with a thermal cycler that has a screw-down or clip-down lid.
  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • Transparent film for fluorescence applications
  • No sticky residue on the plate when seal is removed
  • Temperature range: for film −20 to +120 °C; for foil −80 to +150 °C