Tisk…
Casseroles with handle
Dodavatel: MORGAN ADVANCED MATERIALS HALD
L17/0 L17/000 L17/2 L17/4 L17/4/0
213-5903EA 1730 CZK
213-5903 213-5902 213-5904 213-5905 213-5901
Casseroles with handle
Podnosy/Misky/Kelímky Hrnce
Porcelain, glazed. With spout and porcelain handle. Glazed white inside and outside except for rim.
  • Excellent chemical resistance
  • Heat resistant up to +1000 °C
  • Low thermal expansion
EN 60672-3 Group C100, Type C110
Certifikace: EN 60672-3 Group C100, Type C110