Tisk…
Retort stands
Dodavatel: USBECK Laborgeräte
SET1 SET2
241-5121EA 7910 CZK
241-5121 241-5122
Retort stands
Stojany laboratorní
Stainless steel. Can be used for a wide range of equipment, e.g. stirrers, hotplates etc.
  • Model A: Consists of a base, 10 mm thick, 315×210 mm, and a rod, 800 mm long, 16 mm diameter
  • Model B: Consists of a base, 10 mm thick, 315×210 mm, and a rod, 1000 mm long, 16 mm diameter
  • Robust, firm, corrosion resistant, non slip
  • With three fixed feet and one adjustable
  • Threaded off-centre hole