Tisk…
Odkapávací stojany, nástěnné, LaboPlast®
Dodavatel: BURKLE
9640-1060 9640-2040 9640-2050 9640-2060 9640-1040 9640-1050
149-0193EA 8240 CZK
149-0193 149-1330 149-1331 149-1332 149-0191 149-0192
Odkapávací stojany, nástěnné, LaboPlast®
Stojany Odkapávače
PVC rack with PP pegs.
  • Will not shatter or break
  • Resistant to most chemicals
  • Removable drainage channel with connection for a draining tube hose (for drainage of dripping water into laboratory basin)
This flexible PVC system has stable drainage pegs, available in three lengths (60, 100 and 150 mm) with Ø 6, 10 and 12 mm, that can be repositioned at any time. The draining racks can be adapted to any apparatus which is to be dried.
Informace o dodávce: Rack supplied complete with removable dust and scratch proof protective film, and rods, screws and dowels for wall mounting.