Tisk…
Replication systems, Nunc™
Dodavatel: Thermo Fisher Scientific
250393 250520
732-2706EA 159460 CZK
732-2706 732-2707
Replication systems, Nunc™
Replikátory mikrodestiček a Petriho misek
Anodised aluminium replicators with stainless steel pins, PP copiers. Designed for the replication of DNA libraries, this system is ideal for transferring small volumes of liquid from plate to plate or to a membrane. Pins transfer 1,0 µl (±0,1 µl) of liquid.