Tisk…
Measuring cylinders, tall form, PP
Dodavatel: KARTELL
1075 1077 1078 1079 1080 1081 1082 1094 1082CASE 1094CASE 1080CASE
612-0936EA 227 CZK
612-0936 612-0937 612-0938 612-0939 612-0940 612-0941 612-0942 612-0943 KART1082CASE KART1094CASE KART1080CASE
Measuring cylinders, tall form, PP
Válce Válce odměrné
PP, highly translucent, tall form.
  • Pouring spout, pentagonal base assures excellent stability
  • Non-wetting walls, absolutely no meniscus formation, very good chemical resistance
  • Permanent, embossed graduations
ISO 6706, BS 5404
Certifikace: ISO 6706, BS 5404