Tisk…
Measuring cylinders, tall form, PMP
Dodavatel: KARTELL
1570 1571 1572 1573 1574 1575 1576 1577
612-0951EA 243 CZK
612-0951 612-0952 612-0953 612-0954 612-0955 612-0956 612-0957 612-0958
Measuring cylinders, tall form, PMP
Válce Válce odměrné
PMP (TPX®), transparent, tall form.
  • Pouring spout, pentagon base and moulded transparent graduations
  • No meniscus, chemically non-absorbant, non-wetting surface, excellent chemical resistance
  • Can be used with liquids up to +170 °C
ISO 6706, BS 5404 Part 2
Certifikace: ISO 6706, BS 5404 Part 2