Tisk…
Ion selective half-cell electrodes
Dodavatel: Mettler - Toledo
51340500 51340900 51340800 51340600 51340700 51340400 51340300
662-2800EA 34930 CZK
662-2800 662-2804 662-2807 662-2805 662-2806 662-2801 662-2802
Ion selective half-cell electrodes
Elektrody Elektrody ISE
These ion selective half cells with solid-state membrane allow a quick and easy determination of concentration and activity of a variety of dissolved ions and gases.
  • Modular and versatile ion selective half cells consisting of a universal shaft and a ion specific, exchangeable membrane
  • Conform to RoHS (‘Restriction of Hazardous Substances’ in electrical and electronic equipment) and WEEE (‘Waste Electrical and Electronic Equipment’)
Informace pro objednávání: Membrane kits available separately.