Tisk…
Hydrometers for low and high surface tension
Dodavatel: ALLA ATELIERS
301.070FRG 301.090FRG 301.100FRG 301.110FRG 301.120FRG 301.130FRG 301.140FRG 301.150FRG 301.160FRG 301.170FRG 301.180FRG 301.190FRG 302.060FRG 302.100FRG 302.120FRG 302.140FRG 302.160FRG 302.180FRG
614-0023EA 720 CZK
614-0023 614-0025 614-0026 614-0027 614-0028 614-0029 614-0030 614-0031 614-0032 614-0033 614-0034 614-0035 614-0036 614-0009 614-0038 614-0039 614-0040 614-0041
Hydrometers for low and high surface tension
Hydrometry
Hydrometers for low and high surface tension suitable for general purpose applications.
  • Calibration temperature of 20 °C
  • Reading below the meniscus
  • For low and high surface tension