Tisk…
Biopsy processing/embedding cassettes, Microsette™ I M502
Dodavatel: Simport Scientific
M502-12 M502-5 M502-10 M502-11 M502-2 M502-3 M502-4 M502-6 M502-7 M502-8 M502-9
720-0309EA 13230 CZK
720-0309 720-0954 720-0949 720-0950 720-0951 720-0952 720-0953 720-0955 720-0956 720-0957 720-0958
Biopsy processing/embedding cassettes, Microsette™ I M502
Kazety pro zalévání vzorků Kazety bioptické
Acetal polymer.
  • No need for biopsy pads
  • Large compartment 25×30 mm
  • 2000 holes (Ø 0,26 mm) suitable for needle biopsies
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems