Tisk…
Brýle, kombinéza
Dodavatel: Bollé Safety
COVERSI COVACLEAN
111-0991EA 0 CZK
111-0991 BOLECOVACLEAN
Brýle, kombinéza
Brýle bezpečnostní
Comfortable goggles with adjustable elastic straps.
  • Fit comfortably over most prescription eyewear
  • High flow ventilation with four indirect ventilators
  • Resistant to molten metal and hot solids, dust and liquid aerosols
Certifikace: EN 166